4-Carboxyphenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific
Warning
Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
366390010 366390050
ACRO366390010EA 33.1 EUR
ACRO366390010 ACRO366390050
4-Carboxyphenylboronic acid pinacol ester 97%
4-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
MW: 248.08 g/mol
Melting Pt: 227…232 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863710
CAS Number: 180516-87-4
Order Now
Specification Test Results
Appearance White to cream Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 227 to 232 °C
Titration with NaOH 96.0 to 104.0 %