4-(Carboxymethyl)phenylboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-[(4-Carboxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid, 4-(Carboxymethyl)phenylboronic acid Pinacol ester

718831-250MG 718831-1G
SIAL718831-250MGEA 65.7 EUR
SIAL718831-250MG SIAL718831-1G
4-(Carboxymethyl)phenylboronic acid pinacol ester, Sigma-Aldrich®
4-(Carboxymethyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262.1093 g/mol
MDL Number: MFCD04038755
CAS Number: 797755-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR