4-Iodobenzylamine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: (4-Iodophenyl)methanamine hydrochloride
M-Clarity™ quality level = MQ100
597139-1G 597139-5G
SIAL597139-1GEA 89.3 EUR
SIAL597139-1G SIAL597139-5G
4-Iodobenzylamine hydrochloride, Sigma-Aldrich®
4-Iodobenzylamine hydrochloride
Formula: C₇H₉ClIN
MW: 269.51 g/mol
MDL Number: MFCD04039324
CAS Number: 59528-27-7