Silicon nitride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Silicon(IV) nitride
M-Clarity™ quality level = MQ100
334103-100G 334103-25G
SIAL334103-100GEA 257 EUR
SIAL334103-100G SIAL334103-25G
Silicon nitride, Sigma-Aldrich®
Silicon nitride
Formula: N₄Si₃
MW: 140.28 g/mol
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8