Silicon nitride α-phase, Sigma-Aldrich®
Supplier: Merck
Synonyms: Silicon(IV) nitride
M-Clarity™ quality level = MQ100
325171-250G 325171-50G
SIAL325171-250GEA 280 EUR
SIAL325171-250G SIAL325171-50G
Silicon nitride α-phase, Sigma-Aldrich®
Silicon nitride
Formula: N₄Si₃
MW: 140.28 g/mol
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8