Yttrium(III) 2-ethylhexanoate 10% (w/v) in n-hexane
Supplier: Thermo Fisher Scientific
Danger
43356.AE 43356.AC
43356.AEEA 157 EUR
43356.AE 43356.AC
Yttrium(III) 2-ethylhexanoate 10% (w/v) in n-hexane
Yttrium (III) 2-ethylhexanoate
CAS No.: 103470-68-4 Molecular Formula: C24H45O6Y Formula Weight: 518.16 Physical Form: Liquid UN No.: 1208 MDL No.: MFCD00070441
Formula: C₂₄H₄₅O₆Y
MW: 518.524 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070441
CAS Number: 103470-68-4
UN: 1208
ADR: 3 II