cis-5-Octen-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
cis-5-Octen-1-ol, (Z)-Oct-5-en-1-ol
M-Clarity™ quality level = MQ300
W372218-100G-K
W372218-1KG-K
SIALW372218-100G-KEA
140
EUR
SIALW372218-100G-K
SIALW372218-1KG-K
cis-5-Octen-1-ol, Sigma-Aldrich®
cis-5-Octen-1-ol
Formula:
C₈H₁₆O MW: 128.22 g/mol |
MDL Number:
MFCD00015569 CAS Number: 64275-73-6 EINECS: 264-764-9 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...