Sodium-1-hexadecanesulphonate
Supplier: Molekula
Synonyms: Sodium hexadecane-1-sulphonate, Hexadecane-1-sulphonic acid sodium salt
62080215-100G 62080215-25G
MOLE62080215-100GEA 579 EUR
MOLE62080215-100G MOLE62080215-25G
Sodium-1-hexadecanesulphonate
Sodium 1-hexadecanesulphonate
Formula: C₁₆H₃₃NaSO₃
MW: 328.49 g/mol
MDL Number: MFCD00007528
CAS Number: 15015-81-3
EINECS: 239-100-6