4-Nonylphenylboronic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-N-Nonylphenylboronic acid, 4-n-Nonylbenzeneboronic acid, FAAH/MAGL-IN-4
L17753.03
L17753.06
L17753.03EA
103
EUR
L17753.03
L17753.06
4-Nonylphenylboronic acid ≥98%
4-Nonylphenylboronic acid
Purity: 98+%
CAS No.: 256383-45-6
Molecular Formula: C15H25BO2
Formula Weight: 248.18
Melting Point: 82-85°
Beilstein Reference: 8215118
Formula:
C₁₅H₂₅BO₂ Melting Pt: 82…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093070 CAS Number: 256383-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...