Tetramethylammonium hydroxide 1 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
426318-1L 426318-250ML
SIAL426318-1LEA 172 EUR
SIAL426318-1L SIAL426318-250ML
Tetramethylammonium hydroxide 1 mol/l in aqueous solution, Sigma-Aldrich®
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
Boiling Pt: 110 °C (1013 hPa)
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II