2,2-Dimethoxy-2-phenylacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Irgacure-651
M-Clarity™ quality level = MQ200
196118-250G 196118-50G
SIAL196118-250GEA 319 EUR
SIAL196118-250G SIAL196118-50G
2,2-Dimethoxy-2-phenylacetophenone, Sigma-Aldrich®
2,2-Dimethoxy-2-phenylacetophenone
Formula: C₁₆H₁₆O₃
MW: 256.2964 g/mol
MDL Number: MFCD00008475
CAS Number: 24650-42-8
EINECS: 246-386-6