Digitonin, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Digitonin
M-Clarity™ quality level = MQ200
D141-100MG
D141-500MG
SIALD141-100MGEA
111
EUR
SIALD141-100MG
SIALD141-500MG
Digitonin, Sigma-Aldrich®
Digitonin
Formula:
C₅₆H₉₂O₂₉ MW: 1229.312 g/mol |
MDL Number:
MFCD00077729 CAS Number: 11024-24-1 EINECS: 234-255-6 UN: 3462 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...