α-Terpinene ≥90%
Supplier: Thermo Scientific
Warning
Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene
J62871.AP J62871.AE
J62871.APEA 92 EUR
J62871.AP J62871.AE
α-Terpinene ≥90%
α-Terpinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 173…175 °C
Melting Pt: –48 °C
Density: 0.837
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001534
CAS Number: 99-86-5
EINECS: 202-795-1
UN: 2319
ADR: 3,III
Merck Index: 14,9170