2,4-Dimethyl-1,3-pentadiene ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
H35300.06
H35300.14
H35300.06EA
63.2
EUR
H35300.06
H35300.14
2,4-Dimethyl-1,3-pentadiene ≥98%
2,4-Dimethyl-1,3-pentadiene
Formula:
C₇H₁₂ MW: 96.17 g/mol Boiling Pt: 94…96 °C Melting Pt: –114 °C Density: 0.744 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008903 CAS Number: 1000-86-8 EINECS: 213-677-4 UN: 3295 ADR: 3 II |