Acrylamide 40% aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Prop-2-enamide
J62480.AP J62480.K2
J62480.APEA 119 EUR
J62480.AP J62480.K2
Acrylamide 40% aqueous solution
Acrylamide (monomer)
Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 232
Melting Pt: 84.5
Flash Pt: 138
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 3426
ADR: 6.1 III
Merck Index: 14,129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM