3-Bromo-7-nitro-1H-indazole ≥98%
Supplier: Thermo Scientific
Danger
Synonyms: 3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole
J63480.03 J63480.06
J63480.03EA 101 EUR
J63480.03 J63480.06
3-Bromo-7-nitro-1H-indazole ≥98%
3-Bromo-7-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
Melting Pt: 176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159910
CAS Number: 74209-34-0