4-Amino-1,8-naphthalimide 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Aminonaphthalimide, 4-Aminonaphthalene-1,8-dicarboximide
J64358.MC J64358.03
J64358.MCEA 50 EUR
J64358.MC J64358.03
4-Amino-1,8-naphthalimide 95%
4-Amino-1,8-naphthalimide
Formula: C₁₂H₈N₂O₂
MW: 212.21 g/mol
Boiling Pt: 518
Melting Pt: >360 °C
Density: 1.105
Storage Temperature: Freezer
MDL Number: MFCD00006921
CAS Number: 1742-95-6
EINECS: 217-110-1