Tetranitro blue tetrazolium ≥97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: TNBT, 3,3'-(3,3'-Dimethoxy[1,1'-biphenyl]-4,4'-diyl)bis[2,5-bis(p-nitrophenyl)-2H-tetrazolium] dichloride
J61080.MC J61080.03
J61080.MCEA 92 EUR
J61080.MC J61080.03
Tetranitro blue tetrazolium ≥97%
Tetranitro blue tetrazolium
Purity: 97% CAS No.: 1184-43-6 Molecular Formula: C40H28Cl2N12O10.2C2H6O Formula Weight: 999.78 Physical Form: Powder Appearance: Yellow Beilstein Reference: 3897475 MDL No.: MFCD00036338
Redox indicator for enzymes
Formula: C₄₀H₂₈Cl₂N₁₂O₁₀
MW: 907.64 g/mol
Melting Pt: 165
Storage Temperature: Ambient
MDL Number: MFCD00036338
CAS Number: 1184-43-6
EINECS: 214-665-1