Coomassie® brilliant blue R-250
Supplier: Thermo Scientific
J64297.18EA 98 EUR
J64297.18
Coomassie® brilliant blue R-250
Coomassie® brilliant blue R-250
Formula: C₄₅H₄₄N₃NaO₇S₂
MW: 825,98 g/mol
Melting Pt: 174…180 °C
MDL Number: MFCD00041762
CAS Number: 6104-59-2