Potassium chloride 1 mol/l (1 N) in aqueous solution
Supplier: Cayman Chemical
Synonyms: Potassiumchloride
600213-100 600213-250 600213-500
CAYM600213-100EA 27 EUR
CAYM600213-100 CAYM600213-250 CAYM600213-500
Potassium chloride 1 mol/l (1 N) in aqueous solution
Potassium chloride
Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8