Calcium chloride 1 M in aqueous solution
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
J63122.AD J63122.AE
J63122.ADEA 59.9 EUR
J63122.AD J63122.AE
Calcium chloride 1 M in aqueous solution
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Merck Index: 14,01659