Sodium dodecyl sulphate (SDS) 97% for electrophoresis
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
J64241.14 J64241.22 J64241.36
J64241.14EA 41.7 EUR
J64241.14 J64241.22 J64241.36
Sodium dodecyl sulphate (SDS) 97% for electrophoresis
Sodium dodecyl sulphate (SDS)
Purity: 98%
CAS No.: 151-21-3
Molecular Formula: C12H25NaO4S
Formula Weight: 288.38
Physical Form: Powder
Appearance: White
Melting Point: ca 206°
Solubility: Soluble in water
Beilstein Reference: 3599286
Merck Reference: 14,8636
UN No.: 2926
MDL No.: MFCD00036175
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: ∼206 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 III
Merck Index: 14,8636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM