Hygromycin B
Supplier: Thermo Fisher Scientific
Danger
J60681.MC J60681.MD J60681.03
J60681.MCEA 64 EUR
J60681.MC J60681.MD J60681.03
Hygromycin B
Hygromycin B
Formula: C₂₀H₃₇N₃O₁₃
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1,I
Merck Index: 14,4852