Biebrich scarlet
Supplier: Thermo Scientific
Synonyms: Bonceau BS, C.I. 26905, Acid Red 66, Double Scarlet BSF, Croceine Scarlet, Biebrich Scarlet sodium salt
J65603.14 J65603.22
J65603.14EA 31.1 EUR
J65603.14 J65603.22
Biebrich scarlet
Biebrich Scarlet
Formula: C₂₂H₁₄N₄Na₂O₇S₂
MW: 556.49 g/mol
Melting Pt: 181…188 °C
Flash Pt: 300 °C (572 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003891
CAS Number: 4196-99-0
EINECS: 224-084-5