Rain jacket with detachable softshell jacket, Burma 488A
Supplier: Sioen
Siopor® Ultra
113-7112EA 173 EUR
113-7112 113-7111 113-7114 113-7113 113-7110 113-7109 113-7108 113-7105 113-7104 113-7115 113-7107 113-7106
Rain jacket with detachable softshell jacket, Burma 488A
Workwear
Multifunctional jacket made of Siopor® Ultra fabric (100 % polyester fabric with 100% PU coating). The cold resistant softshell jacket inside is detachable. The mesh lining is fixed and features one inside pocket.
  • Highly breathable, taped seams
  • Round inset sleeves with knitted wind cuffs
  • Straight collar, detachable hood with chin protection
  • Zip closure under flap with touch and close fastening
  • Two inset pockets, Napoleon pocket, Mica pocket
  • Elastic at end of sleeves with narrowing by touch and close fastening, elastic drawstring in waist
The jacket is waterproof and windproof and can therefore be used in all weather conditions.
Back length: 85 cm
Weight: 195 g/m²
Certifikace: EN 343 (3-3)