Sampling fork, Corning® Gosselin™
Supplier: Corning
Corning®
215-2296EA 804 EUR
215-2296
Sampling fork, Corning® Gosselin™
Forks
PS, white.
  • Food contact approved: Raw material is compliant with regulation EC No. 1935/2004
  • Ideal for dairy industries
  • Sterile
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904