Bayonet filters, X-plore® mask
Supplier: Dräger
6738038 6738873 6738872 6738817 6738816 6738775 6738776 6738778 6738039 6738874
111-1046EA 2.57 EUR
111-1046 111-1056 111-1055 111-0770 111-1054 111-1049 111-1052 111-2347 DRAE6738039 DRAE6738874
Bayonet filters, X-plore® mask
Respirators Reusable Respirators Respirator Filters
Range with a Dräger-specific bayonet connection that fits X-plore® 3300/3500 half masks and X-plore® 5500 full face masks.
  • High level of user safety, great fit under face shields and visors
  • Robust gas filter with a blue ABS plastic housing (boiling point >65 °C)
  • White particle filter housing for easy identification of P3 filters
  • Option to upgrade a gas filter to a combination filter by simply adding a P1 or P2 pad or Dräger X-plore® Pure filter
  • Safe, quick and easy bayonet connection with clear guidance for positioning the filter securely
  • Filters inserted top-down for secure positioning
  • Shelf life (from date of production): Gas and combination filters 6 years, particle filters 12 years, pads 3 years
  • Prefilters, to protect combination filters against clogging and blocking
Certifikace: CE certified (EN 14387, EN 143:2000/A1:2006); labelling 'R' (combination filter): Certified for reuse (more than one shift)
Australian Standard AS /NZS 1716 certified (SAI Global)