1,2-Epoxycyclopentane, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Cyclopentene oxide
M-Clarity™ quality level = MQ200
175188-25G
175188-5G
SIAL175188-25GEA
360
EUR
SIAL175188-25G
SIAL175188-5G
1,2-Epoxycyclopentane, Sigma-Aldrich®
1,2-Epoxycyclopentane
Formula:
C₅H₈O MW: 84.12 g/mol |
MDL Number:
MFCD00005161 CAS Number: 285-67-6 EINECS: 206-005-6 |