Lithium aluminum hydride 1 mol/l in diethyl ether, Sigma-Aldrich®
Supplier: Merck
Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate
M-Clarity™ quality level = MQ200
212792-100ML 212792-800ML
SIAL212792-100MLEA 69.4 EUR
SIAL212792-100ML SIAL212792-800ML
Lithium aluminum hydride 1 mol/l in diethyl ether, Sigma-Aldrich®
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3 I
Merck Index: 13,00348