Lithium aluminum hydride 2.2 mol/l, Sigma-Aldrich®
Supplier: Merck
Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate
703508-100ML 703508-800ML
SIAL703508-100MLEA 199 EUR
SIAL703508-100ML SIAL703508-800ML
Lithium aluminum hydride 2.2 mol/l, Sigma-Aldrich®
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
Merck Index: 13,00348