Triethylene glycol bis(chloroformate), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
301477-25G
301477-5G
SIAL301477-25GEA
209
EUR
SIAL301477-25G
SIAL301477-5G
Triethylene glycol bis(chloroformate), Sigma-Aldrich®
Triethylene glycol bis(chloroformate)
Formula:
C₈H₁₂Cl₂O₆ |
MDL Number:
MFCD00011536 CAS Number: 17134-17-7 UN: 3277 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...