2,2,2-Trifluoroethyl trichloromethanesulphonate ≥94%
Supplier: Thermo Fisher Scientific
|
Danger
|
L08635.06
L08635.14
L08635.06EA
118
EUR
L08635.06
L08635.14
2,2,2-Trifluoroethyl trichloromethanesulphonate ≥94%
2,2,2-Trifluoroethyl trichloromethanesulphonate
Purity: 94%
CAS No.: 23199-56-6
Molecular Formula: Cl3CSO2OCH2CF3
Formula Weight: 281.47
Refractive Index: 1.4220
Odour: Acrid
Boiling Point: 84-86°/20mm
Beilstein Reference: 1108366
UN No.: 3265
MDL No.: MFCD00042400
Formula:
C₃H₂Cl₃F₃O₃S Boiling Pt: 84…86 °C (20 mmHg) Melting Pt: –70 °C Density: 1.714 Storage Temperature: Ambient |
MDL Number:
MFCD00042400 CAS Number: 23199-56-6 EINECS: 245-484-6 UN: 3265 ADR: 8,II |