Ethyl acetopyruvate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethyl 2,4-dioxopentanoate, Ethyl 2,4-dioxovalerate

M-Clarity™ quality level = MQ200

232564-25G 232564-5G
SIAL232564-25GEA 69.6 EUR
SIAL232564-25G SIAL232564-5G
Ethyl acetopyruvate, Sigma-Aldrich®
Ethyl acetopyruvate
Formula: C₇H₁₀O₄
MW: 158.15 g/mol
MDL Number: MFCD00009124
CAS Number: 615-79-2
EINECS: 210-447-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR