L(+)-Diisopropyl tartrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Diisopropyl (2R,3R)-2,3-dihydroxysuccinate, L-(+)-Tartaric acid diisopropyl ester

M-Clarity™ quality level = MQ100

229180-100G 229180-25G
SIAL229180-100GEA 72.9 EUR
SIAL229180-100G SIAL229180-25G
L(+)-Diisopropyl tartrate, Sigma-Aldrich®
L(+)-Diisopropyl tartrate
Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
MDL Number: MFCD00064450
CAS Number: 2217-15-4
EINECS: 218-709-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR