N-Methyliminodiacetic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2,2'-(Methylazanediyl)diacetic acid
M-Clarity™ quality level = MQ100
M51008-25G
M51008-5G
SIALM51008-25GEA
144
EUR
SIALM51008-25G
SIALM51008-5G
N-Methyliminodiacetic acid, Sigma-Aldrich®
N-Methyliminodiacetic acid
Formula:
C₅H₉NO₄ MW: 147.13 g/mol |
MDL Number:
MFCD00004284 CAS Number: 4408-64-4 EINECS: 224-557-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...