N-Methyliminodiacetic acid disodium salt, Sigma-Aldrich®

Supplier: Merck

705012-1G 705012-5G
SIAL705012-1GEA 41 EUR
SIAL705012-1G SIAL705012-5G
N-Methyliminodiacetic acid disodium salt, Sigma-Aldrich®
N-Methyliminodiacetic acid disodium salt
Formula: C₅H₇NNa₂O₄
MDL Number: MFCD12546042
CAS Number: 71160-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR