3-Aminopropionitrile fumarate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A3134-10G
A3134-25G
A3134-5G
SIALA3134-10GEA
152
EUR
SIALA3134-10G
SIALA3134-25G
SIALA3134-5G
3-Aminopropionitrile fumarate, Sigma-Aldrich®
3-Aminopropionitrile fumarate
Formula:
C₁₀H₁₆N₄O₄ MW: 256.26 g/mol |
MDL Number:
MFCD00013142 CAS Number: 2079-89-2 EINECS: 218-208-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...