Methyl vinyl sulphone, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

247197-25G 247197-5G
SIAL247197-25GEA 147 EUR
SIAL247197-25G SIAL247197-5G
Methyl vinyl sulphone, Sigma-Aldrich®
Methyl vinyl sulphone
Formula: C₃H₆O₂S
MW: 106.15 g/mol
MDL Number: MFCD00007567
CAS Number: 3680-02-2
EINECS: 222-958-0
UN: 2810
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR