Borane trimethylamine complex, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

178985-25G 178985-5G 178985-100G
SIAL178985-25GEA 99.6 EUR
SIAL178985-25G SIAL178985-5G SIAL178985-100G
Borane trimethylamine complex, Sigma-Aldrich®
Borane trimethylamine complex
Formula: C₃H₁₂BN
MW: 72.95 g/mol
MDL Number: MFCD00012420
CAS Number: 75-22-9
EINECS: 200-850-4
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR