[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: [1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd(dppf)Cl2, 1,1’-Bis(diphenylphosphino)ferrocene palladium(II)dichloride
M-Clarity™ quality level = MQ200
697230-1G 697230-5G
SIAL697230-1GEA 70.1 EUR
SIAL697230-1G SIAL697230-5G
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride, Sigma-Aldrich®
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) chloride
Formula: C₃₄H₂₈Cl₂P₂FePd
MDL Number: MFCD00015757
CAS Number: 72287-26-4
EINECS: 460-040-3