(Trimethylsilyl)diazomethane 2 M hexanes
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Trimethylsilyldiazomethane, TMS-Diazomethane
385330050 385330250 385331000 385335000
ACRO385330050EA 77.5 EUR
ACRO385330050 ACRO385330250 ACRO385331000 ACRO385335000
(Trimethylsilyl)diazomethane 2 M hexanes
(Trimethylsilyl)diazomethane
Formula: C₄H₁₀N₂Si
MW: 114.22 g/mol
Density: 0.675 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00053946
CAS Number: 18107-18-1
UN: 1992
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear yellow Solution
Strength 1.9 to 2.4 M