Folic acid cell culture reagent
Supplier: Thermo Scientific
Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
J62937.06 J62937.14 J62937.22
J62937.06EA 38.6 EUR
J62937.06 J62937.14 J62937.22
Folic acid cell culture reagent
Folic acid
CAS No.: 59-30-3 Molecular Formula: C19H19N7O6 Formula Weight: 441.41 Physical Form: Powder Appearance: Yellow to orange Beilstein Reference: 100781 Merck Reference: 14,4221 MDL No.: MFCD00079305
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells
Formula: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0
Merck Index: 14,4221