D-(-)-Luciferin sodium salt monohydrate ≥98%, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

L6882-.2MG L6882-100MG L6882-10MG L6882-2MG L6882-50MG L6882-5MG
SIALL6882-.2MGEA 48.2 EUR
SIALL6882-.2MG SIALL6882-100MG SIALL6882-10MG SIALL6882-2MG SIALL6882-50MG SIALL6882-5MG
D-(-)-Luciferin sodium salt monohydrate ≥98%, Sigma-Aldrich®
D(-)-Luciferin sodium salt
Formula: C₁₁H₇N₂NaO₃S₂·H₂O
MW: 320.32 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11865368
CAS Number: 103404-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR