Lansoprazole

Supplier: Molekula

MOLEM89022451EA 0 EUR
MOLEM89022451
Lansoprazole
Lansoprazole
Formula: C₁₆H₁₄F₃N₃O₂S
MW: 369,36 g/mol
CAS Number: 103577-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR