Theophyllin
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline
J60203.5H J60203.RX
J60203.5HEA 87.6 EUR
J60203.5H J60203.RX
Theophyllin
Theophyllin
Formula: C₇H₈N₄O₂
MW: 180.16 g/mol
Melting Pt: 270…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00079619
CAS Number: 58-55-9
EINECS: 200-385-7
UN: 2811
ADR: 6.1 III
Merck Index: 14,9285