Theophylline-7-acetic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)acetic acid
A11473.14
A11473.22
A11473.36
A11473.14EA
115
EUR
A11473.14
A11473.22
A11473.36
Theophylline-7-acetic acid ≥98%
Theophylline-7-acetic acid
Purity: 98%
CAS No.: 652-37-9
Molecular Formula: C9H10N4O4
Formula Weight: 238.20
Melting Point: 268-272°
Beilstein Reference: 279221
Merck Reference: 14,24
MDL No.: MFCD00022832
Formula:
C₉H₁₀N₄O₄ Melting Pt: 268…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022832 CAS Number: 652-37-9 EINECS: 211-490-2 Merck Index: 14,24 |