Forskolin, Sigma-Aldrich®
Supplier: Merck
Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
M-Clarity™ quality level = MQ200
F3917-10MG F3917-25MG
SIALF3917-10MGEA 303 EUR
SIALF3917-10MG SIALF3917-25MG
Forskolin, Sigma-Aldrich®
Forskolin
Formula: C₂₂H₃₄O₇
MW: 410.51 g/mol
MDL Number: MFCD00082317
CAS Number: 66575-29-9
EINECS: 266-410-9