(R)-1-Boc-3-Hydroxypiperidine ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate
H52556.03
H52556.06
H52556.03EA
77.5
EUR
H52556.03
H52556.06
(R)-1-Boc-3-Hydroxypiperidine ≥97%
(R)-1-Boc-3-hydroxypiperidine
Purity: 97%
CAS No.: 143900-43-0
Molecular Formula: C10H19NO3
Formula Weight: 201.27
Flash Point: 104°(219°F)
Melting Point: 46-50°
Optical Rotation: -23° (c=5 in methanol)
MDL No.: MFCD04115306
Formula:
C₁₀H₁₉NO₃ Melting Pt: 46…50 °C Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04115306 CAS Number: 143900-43-0 |