Allantoin 98%
Supplier: Thermo Scientific
Synonyms: 1-(2,5-Dioxoimidazolidin-4-yl)urea
102841000 102845000
ACRO102841000EA 33.1 EUR
ACRO102841000 ACRO102845000
Allantoin 98%
Allantoin
Formula: C₄H₆N₄O₃
MW: 158.12 g/mol
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005260
CAS Number: 97-59-6
EINECS: 202-592-8
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Titration with HClO4 ≥98.5 %