3,4-(Methylenedioxy)phenylboronic acid 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3,4-(Methylenedioxy)benzeneboronic acid, Benzo[d][1,3]dioxol-5-ylboronic acid
359580010 359580050
ACRO359580010EA 39.3 EUR
ACRO359580010 ACRO359580050
3,4-(Methylenedioxy)phenylboronic acid 98%
3,4-(Methylenedioxy)phenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01009695
CAS Number: 94839-07-3
Order Now
Specification Test Results
Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Titration with NaOH 97.5 to 106.0 %